Assay Details
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600748.3 | XP_005600805.2 | 1627 | 90 | ||
XM_005600750.3 | XP_005600807.2 | 1590 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
