Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501559.5 | XP_001501609.1 | 258 | 68 | ||
XM_023620676.1 | XP_023476444.1 | 259 | 68 | |||
XM_023620677.1 | XP_023476445.1 | 260 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
