Assay Details
Gene Symbol: EIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731767.2 | XP_014587253.2 | 410 | 152 | ||
XM_023620698.1 | XP_023476466.1 | 475 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
