Assay Details
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620795.1 | XP_023476563.1 | 649 | 68 | ||
XM_023620796.1 | XP_023476564.1 | 673 | 68 | |||
XM_023620801.1 | XP_023476569.1 | 496 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
