Assay Details
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620852.1 | XP_023476620.1 | 569 | 72 | ||
XM_023620853.1 | XP_023476621.1 | 2028 | 72 | |||
XM_023620854.1 | XP_023476622.1 | 1050 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
