Assay Details
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600899.3 | XP_005600956.1 | 457 | 98 | ||
XM_023620855.1 | XP_023476623.1 | 388 | 98 | |||
GenBank mRNA | JL619370.1 | - | 2 - 3 | 110 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
