Assay Details
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917735.5 | XP_001917770.4 | 1429 | 67 | ||
XM_023620865.1 | XP_023476633.1 | 1484 | 67 | |||
XM_023620866.1 | XP_023476634.1 | 2039 | 67 | |||
XM_023620867.1 | XP_023476635.1 | 1564 | 67 | |||
XM_023620868.1 | XP_023476636.1 | 2118 | 67 | |||
XM_023620869.1 | XP_023476637.1 | 1527 | 67 | |||
XM_023620870.1 | XP_023476638.1 | 2081 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
