Assay Details
Gene Symbol: MRPS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495342.5 | XP_001495392.3 | 912 | 78 | ||
XM_014731892.2 | XP_014587378.2 | 1023 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
