Assay Details
Gene Symbol: SGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489821.5 | XP_001489871.1 | 555 | 72 | ||
XM_014731968.2 | XP_014587454.1 | 514 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
