Assay Details
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642813.1 | XP_023498581.1 | 512 | 72 | ||
XM_023642814.1 | XP_023498582.1 | 512 | 72 | |||
XM_023642815.1 | XP_023498583.1 | 512 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
