Assay Details
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740623.2 | XP_014596109.1 | 397 | 72 | ||
XM_023642820.1 | XP_023498588.1 | 725 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
