Assay Details
Gene Symbol: TUBA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489082.5 | XP_001489132.1 | 350 | 70 | ||
XM_023642819.1 | XP_023498587.1 | 335 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
