Assay Details
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643021.1 | XP_023498789.1 | 1442 | 95 | ||
XM_023643022.1 | XP_023498790.1 | 1439 | 95 | |||
XM_023643023.1 | XP_023498791.1 | 1443 | 95 | |||
XM_023643024.1 | XP_023498792.1 | 1347 | 95 | |||
XM_023643025.1 | XP_023498793.1 | 1344 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
