Assay Details
Gene Symbol: CSRNP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496003.4 | XP_001496053.1 | 1245 | 72 | ||
XM_023623064.1 | XP_023478832.1 | 511 | 72 | |||
XM_023623065.1 | XP_023478833.1 | 1527 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
