Assay Details
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601623.3 | XP_005601680.1 | 626 | 233 | ||
XM_005601624.3 | XP_005601681.1 | 527 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
