Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501963.6 | XP_001502013.1 | 241 | 94 | ||
XM_005601634.3 | XP_005601691.1 | 362 | 94 | |||
XM_005601635.3 | XP_005601692.1 | 398 | 94 | |||
XM_023622190.1 | XP_023477958.1 | 493 | 94 | |||
XM_023622192.1 | XP_023477960.1 | 493 | 94 | |||
XM_023622193.1 | XP_023477961.1 | 493 | 94 | |||
XM_023622194.1 | XP_023477962.1 | 411 | 94 | |||
XM_023622195.1 | XP_023477963.1 | 567 | 94 | |||
XM_023622196.1 | XP_023477964.1 | 396 | 94 | |||
XM_023622197.1 | XP_023477965.1 | 167 | 94 | |||
XM_023622198.1 | XP_023477966.1 | 681 | 94 | |||
XM_023622199.1 | XP_023477967.1 | 288 | 94 | |||
XM_023622200.1 | XP_023477968.1 | 493 | 94 | |||
XM_023622201.1 | XP_023477969.1 | 493 | 94 | |||
XM_023622202.1 | XP_023477970.1 | 493 | 94 | |||
XM_023622203.1 | XP_023477971.1 | 493 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
