Assay Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622403.1 | XP_023478171.1 | 2867 | 69 | ||
XM_023622404.1 | XP_023478172.1 | 2867 | 69 | |||
XM_023622405.1 | XP_023478173.1 | 2798 | 69 | |||
XM_023622406.1 | XP_023478174.1 | 2711 | 69 | |||
XM_023622407.1 | XP_023478175.1 | 2411 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
