Assay Details
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732716.2 | XP_014588202.1 | 1418 | 85 | ||
XM_023623237.1 | XP_023479005.1 | 453 | 85 | |||
XM_023623238.1 | XP_023479006.1 | 517 | 85 | |||
XM_023623239.1 | XP_023479007.1 | 534 | 85 | |||
XM_023623240.1 | XP_023479008.1 | 461 | 85 | |||
XM_023623241.1 | XP_023479009.1 | 514 | 85 | |||
XM_023623242.1 | XP_023479010.1 | 421 | 85 | |||
XM_023623244.1 | XP_023479012.1 | 389 | 85 | |||
XM_023623245.1 | XP_023479013.1 | 450 | 85 | |||
XM_023623247.1 | XP_023479015.1 | 1339 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023623244.1, XM_023623245.1, XM_023623247.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
