Assay Details
Gene Symbol: ZMAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494925.5 | XP_001494975.1 | 775 | 58 | ||
XM_005601818.3 | XP_005601875.1 | 766 | 58 | |||
XM_023623331.1 | XP_023479099.1 | 1847 | 58 | |||
XM_023623332.1 | XP_023479100.1 | 1961 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
