Assay Details
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498257.4 | XP_001498307.1 | 1880 | 61 | ||
XM_023623433.1 | XP_023479201.1 | 3214 | 61 | |||
XM_023623434.1 | XP_023479202.1 | 3214 | 61 | |||
XM_023623435.1 | XP_023479203.1 | 3214 | 61 | |||
XM_023623436.1 | XP_023479204.1 | 3214 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498257.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
