Assay Details
Gene Symbol: FGF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500259.6 | XP_001500309.1 | 882 | 62 | ||
XM_003363362.4 | XP_003363410.1 | 358 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
