Assay Details
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500837.4 | XP_001500887.1 | 522 | 70 | ||
XM_005601895.3 | XP_005601952.1 | 479 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
