Assay Details
Gene Symbol: ILDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916842.5 | XP_001916877.4 | 587 | 67 | ||
XM_014732857.2 | XP_014588343.2 | 591 | 67 | |||
XM_014732858.2 | XP_014588344.2 | 595 | 67 | |||
XM_014732859.2 | XP_014588345.2 | 593 | 67 | |||
XM_014732860.2 | XP_014588346.2 | 592 | 67 | |||
XM_023623664.1 | XP_023479432.1 | 588 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
