Assay Details
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500767.4 | XP_001500817.1 | 959 | 60 | ||
XM_023623714.1 | XP_023479482.1 | 926 | 60 | |||
XM_023623715.1 | XP_023479483.1 | 963 | 60 | |||
XM_023623717.1 | XP_023479485.1 | 1664 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
