Assay Details
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601966.3 | XP_005602023.1 | 360 | 88 | ||
XM_014732874.2 | XP_014588360.1 | 470 | 88 | |||
XM_023623720.1 | XP_023479488.1 | 354 | 88 | |||
XM_023623721.1 | XP_023479489.1 | 471 | 88 | |||
XM_023623722.1 | XP_023479490.1 | 293 | 88 | |||
XM_023623723.1 | XP_023479491.1 | 608 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023623722.1, XM_023623723.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
