Assay Details
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601973.3 | XP_005602030.1 | 566 | 108 | ||
XM_005601974.3 | XP_005602031.1 | 517 | 108 | |||
XM_023623745.1 | XP_023479513.1 | 456 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
