Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501316.4 | XP_001501366.1 | 387 | 78 | ||
XM_005602003.3 | XP_005602060.1 | 390 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
