Assay Details
Gene Symbol: ABHD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503160.4 | XP_001503210.1 | 620 | 71 | ||
GenBank mRNA | JL622625.1 | - | 4 - 5 | 532 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
