Assay Details
Gene Symbol: NSUN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623990.1 | XP_023479758.1 | 982 | 56 | ||
XM_023623991.1 | XP_023479759.1 | 905 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
