Assay Details
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488028.5 | XP_001488078.1 | 1340 | 88 | ||
XM_005607020.3 | XP_005607077.1 | 891 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
