Assay Details
Gene Symbol: AGBL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737468.2 | XP_014592954.2 | 189 | 65 | ||
XM_023636827.1 | XP_023492595.1 | 188 | 65 | |||
XM_023636828.1 | XP_023492596.1 | 189 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014737468.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
