Assay Details
Gene Symbol: MOB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607077.3 | XP_005607134.1 | 609 | 70 | ||
XM_005607078.3 | XP_005607135.1 | 640 | 70 | |||
XM_005607079.3 | XP_005607136.1 | 621 | 70 | |||
XM_023630566.1 | XP_023486334.1 | 610 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
