Assay Details
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495067.5 | XP_001495117.1 | 1087 | 67 | ||
XM_005607083.3 | XP_005607140.1 | 1479 | 67 | |||
XM_023630633.1 | XP_023486401.1 | 1032 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
