Assay Details
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496034.5 | XP_001496084.1 | 539 | 80 | ||
XM_005607090.3 | XP_005607147.1 | 538 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
