Assay Details
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497177.6 | XP_001497227.1 | 987 | 65 | ||
XM_005607113.3 | XP_005607170.1 | 987 | 65 | |||
XM_005607114.3 | XP_005607171.1 | 987 | 65 | |||
XM_014737338.2 | XP_014592824.1 | 570 | 65 | |||
XM_023631440.1 | XP_023487208.1 | 987 | 65 | |||
XM_023631441.1 | XP_023487209.1 | 987 | 65 | |||
XM_023631466.1 | XP_023487234.1 | 334 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
