Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916449.5 | XP_001916484.1 | 958 | 87 | ||
XM_023632614.1 | XP_023488382.1 | 553 | 87 | |||
XM_023632620.1 | XP_023488388.1 | 399 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
