Assay Details
Gene Symbol: MRPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503613.5 | XP_001503663.1 | 753 | 71 | ||
GenBank mRNA | JL625803.1 | - | 1 - 2 | 139 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
