Assay Details
Gene Symbol: ZMYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499380.5 | XP_001499430.4 | 1299 | 102 | ||
XM_005607214.3 | XP_005607271.2 | 1254 | 102 | |||
XM_005607215.3 | XP_005607272.2 | 1103 | 102 | |||
XM_005607216.3 | XP_005607273.2 | 1030 | 102 | |||
XM_014737480.2 | XP_014592966.2 | 1524 | 102 | |||
XM_014737481.2 | XP_014592967.2 | 576 | 102 | |||
XM_023633686.1 | XP_023489454.1 | 1086 | 102 | |||
XM_023633707.1 | XP_023489475.1 | 288 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
