Assay Details
Gene Symbol: YARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634189.1 | XP_023489957.1 | 1197 | 59 | ||
XM_023634193.1 | XP_023489961.1 | 1160 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
