Assay Details
Gene Symbol: EIF3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503817.6 | XP_001503867.5 | 647 | 61 | ||
XM_014737311.2 | XP_014592797.1 | 547 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
