Assay Details
Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503980.5 | XP_001504030.1 | 822 | 72 | ||
XM_005607272.3 | XP_005607329.1 | 820 | 72 | |||
XM_005607273.3 | XP_005607330.1 | 703 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
