Assay Details
Gene Symbol: PPP1R8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504008.5 | XP_001504058.3 | 153 | 84 | ||
XM_023635162.1 | XP_023490930.1 | 153 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023635162.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
