Assay Details
Gene Symbol: PINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737247.2 | XP_014592733.2 | 447 | 63 | ||
XM_023635588.1 | XP_023491356.1 | 449 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
