Assay Details
Gene Symbol: PLA2G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607394.3 | XP_005607451.1 | 606 | 63 | ||
XM_023635597.1 | XP_023491365.1 | 280 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
