Assay Details
Gene Symbol: OTUD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501720.5 | XP_001501770.3 | 347 | 59 | ||
XM_023635599.1 | XP_023491367.1 | 348 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
