Assay Details
Gene Symbol: RIMKLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643100.1 | XP_023498868.1 | 1050 | 65 | ||
XM_023643101.1 | XP_023498869.1 | 997 | 65 | |||
XM_023643102.1 | XP_023498870.1 | 829 | 65 | |||
XM_023643103.1 | XP_023498871.1 | 784 | 65 | |||
XM_023643104.1 | XP_023498872.1 | 786 | 65 | |||
XM_023643105.1 | XP_023498873.1 | 1050 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
