Assay Details
Gene Symbol: ERP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501766.4 | XP_001501816.1 | 1134 | 89 | ||
XM_023643171.1 | XP_023498939.1 | 1134 | 89 | |||
XM_023643172.1 | XP_023498940.1 | 1134 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
