Assay Details
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501778.4 | XP_001501828.1 | 405 | 85 | ||
XM_005610958.3 | XP_005611015.1 | 388 | 85 | |||
XM_023643173.1 | XP_023498941.1 | 321 | 85 | |||
GenBank mRNA | JL617600.1 | - | 3 - 4 | 342 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
