Assay Details
Gene Symbol: DHRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490729.6 | XP_001490779.1 | 1285 | 65 | ||
XM_023635755.1 | XP_023491523.1 | 547 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
