Assay Details
Gene Symbol: GABRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635949.1 | XP_023491717.1 | 1296 | 76 | ||
XM_023635950.1 | XP_023491718.1 | 1268 | 76 | |||
XM_023635951.1 | XP_023491719.1 | 1247 | 76 | |||
XM_023635952.1 | XP_023491720.1 | 1228 | 76 | |||
XM_023635953.1 | XP_023491721.1 | 1180 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
